Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethyl)benzoic acid, 97+%

$47.91 - $153.69

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzoic acid
CAS 654-99-9
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD00083521
Beilstein 2618195
Synonym alpha,alpha,alpha,5-Tetrafluoro-o-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2314903

  SDS
alfa aesar™
B2314903
1g Each for $47.91

AAB2314906

  SDS
alfa aesar™
B2314906
5g Each for $153.69
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzoic acid
Assay ≥97%
Molecular Formula C8H4F4O2
MDL Number MFCD00083521
CAS 654-99-9
Beilstein 2618195
Synonym alpha,alpha,alpha,5-Tetrafluoro-o-toluic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 80° to 83°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.