Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethyl)benzaldehyde, 97%

$109.85 - $442.99

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzaldehyde
CAS 90381-08-1
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00236276
Synonym alpha,alpha,alpha,5-Tetrafluoro-o-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1990103

  SDS
alfa aesar™
L1990103
1g Each for $109.85

AAL1990106

  SDS
alfa aesar™
L1990106
5g Each for $442.99
Description & Specifications

Specifications

Formula Weight 192.11
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzaldehyde
CAS 90381-08-1
Molecular Formula C8H4F4O
MDL Number MFCD00236276
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym alpha,alpha,alpha,5-Tetrafluoro-o-tolualdehyde
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.