Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-(trifluoromethyl)acetophenone, 97%

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-(trifluoromethyl)acetophenone
Molecular Formula C9H6F4O
Formula Weight 206.14
MDL Number MFCD00236274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3217103

  SDS
alfa aesar™
H3217103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3217106

  SDS
alfa aesar™
H3217106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.14
Quantity 1g
Molecular Formula C9H6F4O
Chemical Name or Material 5'-Fluoro-2'-(trifluoromethyl)acetophenone
MDL Number MFCD00236274
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.