Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethoxy)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzoyl chloride
Molecular Formula C8H3ClF4O2
Formula Weight 242.55
MDL Number MFCD11519334
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3280403

  SDS
alfa aesar™
H3280403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32804MD

  SDS
alfa aesar™
H32804MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.55
Assay 97%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C8H3ClF4O2
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzoyl chloride
UN Number UN3265
MDL Number MFCD11519334
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.