Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethoxy)benzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzaldehyde
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD11519331
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3295606

  SDS
alfa aesar™
H3295606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3295603

  SDS
alfa aesar™
H3295603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O2
MDL Number MFCD11519331
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 5g

Applications
It is an organic fluorinated building block. It participates in the synthesis of diphenylthioethers. It also participates in the preparation of 1-cyano derivative of nilutamide.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.