Please login to your online account to display your discounted pricing

5-Fluoro-2-nitrotoluene 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-nitrotoluene
Name Note 96%
CAS 446-33-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H6FNO2
Linear Formula CH3C6H3(NO2)F
Formula Weight 155.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119530500

  SDS
thermo fisher scientific
119530500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.13
Physical Form Liquid After Melting
Assay ≥95%
Quantity 50g
Color Clear yellow to yellow-green
Beilstein 05, II, 250
EINECS Number 207-164-4
Density 1.2700g/mL
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5260 to 1.5300 (20°C, 589nm)
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 5-Fluoro-2-nitrotoluene
Assay Percent Range 96%
CAS 446-33-3
Linear Formula CH3C6H3(NO2)F
Molecular Formula C7H6FNO2
MDL Number MFCD00007283
Name Note 96%