Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-nitrobenzotrifluoride, 98+%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-nitrobenzotrifluoride
CAS 393-09-9
Molecular Formula C7H3F4NO2
Formula Weight 209.1
MDL Number MFCD00039741
UN Number UN2811
Beilstein 3304470
Synonym 4-Fluoro-2-(trifluoromethyl)nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1878006

 
alfa aesar™
A1878006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1878014

 
alfa aesar™
A1878014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.1
Chemical Name or Material 5-Fluoro-2-nitrobenzotrifluoride
Assay ≥98%
Molecular Formula C7H3F4NO2
MDL Number MFCD00039741
CAS 393-09-9
UN Number UN2811
Beilstein 3304470
Synonym 4-Fluoro-2-(trifluoromethyl)nitrobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point 88°C (190°F)
Boiling Point 87° to 88°C (2mmHg)
Melting Point 22° to 24°C
Quantity 5g

Applications
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.