Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-nitrobenzoic acid, 97+%

$47.86 - $163.16

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-nitrobenzoic acid
CAS 320-98-9
Molecular Formula C7H4FNO4
Formula Weight 185.11
MDL Number MFCD00055635
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2313303

  SDS
alfa aesar™
B2313303
1g Each for $47.86

AAB2313306

  SDS
alfa aesar™
B2313306
5g Each for $163.16
Description & Specifications

Specifications

Formula Weight 185.11
Chemical Name or Material 5-Fluoro-2-nitrobenzoic acid
Assay ≥97%
Molecular Formula C7H4FNO4
MDL Number MFCD00055635
CAS 320-98-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 134° to 135°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.