missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%

$243.03 - $722.72

Chemical Identifiers

CAS 925207-14-3
Molecular Formula C12H15BFNO4
Molecular Weight (g/mol) 267.063
MDL Number MFCD19288891
InChI Key JCPLXROABVMFQO-UHFFFAOYSA-N
Synonym 2-5-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-fluoro-2-nitrophenylboronic acid pinacol ester
PubChem CID 15946129
IUPAC Name 2-(5-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62813MD
View Documents
Alfa Aesar™
H62813MD
250mg
Each for $243.03
 
AAH6281303
View Documents
Alfa Aesar™
H6281303
1g
Each for $722.72
 
Description

Description

Specifications

Chemical Identifiers

925207-14-3
267.063
JCPLXROABVMFQO-UHFFFAOYSA-N
15946129
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)[N+](=O)[O-]
C12H15BFNO4
MFCD19288891
2-5-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-fluoro-2-nitrophenylboronic acid pinacol ester
2-(5-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

250mg
C12H15BFNO4
2-5-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-fluoro-2-nitrophenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)[N+](=O)[O-]
267.063
267.06
96%
925207-14-3
MFCD19288891
JCPLXROABVMFQO-UHFFFAOYSA-N
2-(5-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
15946129
Solid
5-Fluoro-2-nitrobenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS