Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-nitrobenzaldehyde, 98%

$114.36 - $424.84

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-nitrobenzaldehyde
CAS 395-81-3
Molecular Formula C7H4FNO3
Formula Weight 169.11
MDL Number MFCD00153175
Beilstein 1869010
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1932703

 
alfa aesar™
L1932703
1g Each for $114.36

AAL1932706

 
alfa aesar™
L1932706
5g Each for $424.84
Description & Specifications

Specifications

Formula Weight 169.11
Assay 98%
CAS 395-81-3
Beilstein 1869010
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 153°C (23mmHg)
Chemical Name or Material 5-Fluoro-2-nitrobenzaldehyde
Molecular Formula C7H4FNO3
MDL Number MFCD00153175
Melting Point 93° to 96°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.