Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylphenylhydrazine hydrochloride, 97%

$81.43 - $81.43

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylphenylhydrazine hydrochloride
CAS 325-50-8
Molecular Formula C7H9FN2·HCl
Formula Weight 176.62
MDL Number MFCD00053032
Beilstein 3696216
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0936203

  SDS
alfa aesar™
L0936203
1g Each for $81.43
Description & Specifications

Specifications

Formula Weight 176.62
Chemical Name or Material 5-Fluoro-2-methylphenylhydrazine hydrochloride
Assay 97%
Molecular Formula C7H9FN2·HCl
MDL Number MFCD00053032
Beilstein 3696216
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Melting Point ≈197°C
Quantity 1g
CAS 325-50-8
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.