Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylphenol, 98%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylphenol
CAS 452-85-7
Molecular Formula C7H7FO
Formula Weight 126.13
MDL Number MFCD00673009
UN Number UN3265
Beilstein 3234863
Synonym 5-Fluoro-o-cresol; 4-Fluoro-2-hydroxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2560603

  SDS
alfa aesar™
B2560603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2560606

  SDS
alfa aesar™
B2560606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.13
Chemical Name or Material 5-Fluoro-2-methylphenol
Assay 98%
Molecular Formula C7H7FO
MDL Number MFCD00673009
CAS 452-85-7
UN Number UN3265
Beilstein 3234863
Synonym 5-Fluoro-o-cresol; 4-Fluoro-2-hydroxytoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Odor Phenol-like
Boiling Point 66°C (4mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.