Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylbenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylbenzyl bromide
CAS 261951-71-7
Molecular Formula C8H8BrF
Formula Weight 203.06
MDL Number MFCD01631528
UN Number UN3265
Synonym 2-Bromomethyl-4-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2685303

  SDS
alfa aesar™
H2685303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2685306

  SDS
alfa aesar™
H2685306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.06
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Synonym 2-Bromomethyl-4-fluorotoluene
UN Number UN3265
Chemical Name or Material 5-Fluoro-2-methylbenzyl bromide
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Assay 97%
Molecular Formula C8H8BrF
MDL Number MFCD01631528
CAS 261951-71-7
Boiling Point 86°C (3mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.