Please login to your online account to display your discounted pricing

5-Fluoro-2-methylbenzoic acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  310420050

 View more versions of this product

Catalog No. AC310420050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 129° to 132°C
Color Beige to light gray
Quantity 5g
Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
CAS 33184-16-6
CAS Min % 98.5
CAS Max % 100.0
Physical Form Beige to light gray crystalline powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
Formula Weight 154.14g/mol
MDL Number MFCD00042294
Synonym 5-Fluoro-o-toluic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
CAS 33184-16-6
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
Formula Weight 154.14g/mol
MDL Number MFCD00042294
Synonym 5-Fluoro-o-toluic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents