Please login to your online account to display your discounted pricing

5-Fluoro-2-methylbenzoic acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC310420010

 View more versions of this product

Catalog No. AC310420010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 154.14
Quantity 1g
Color Beige to light gray
Melting Point 129° to 132°C
CAS Max % 100.0
Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
MDL Number MFCD00042294
CAS 33184-16-6
Synonym 5-Fluoro-o-toluic acid
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
CAS 33184-16-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
Formula Weight 154.14
MDL Number MFCD00042294
Synonym 5-Fluoro-o-toluic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents