Please login to your online account to display your discounted pricing

5-Fluoro-2-methylbenzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
CAS 33184-16-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
Formula Weight 154.14
MDL Number MFCD00042294
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC310420010

  SDS
thermo fisher scientific
AC310420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 154.14
Physical Form Crystalline Powder
Quantity 1g
Color Beige to light gray
Melting Point 129° to 132°C
CAS Max % 100.0
Packaging Glass bottle
Linear Formula FC6H3(CH3)COOH
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
MDL Number MFCD00042294
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
CAS Min % 98.5
Molecular Formula C8H7FO2
Synonym 5-Fluoro-o-toluic acid
Infrared Spectrum Authentic
CAS 33184-16-6