missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylbenzoic acid, 99%

$27.76 - $479.59

Chemical Identifiers

CAS 33184-16-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00042294
InChI Key JVBLXLBINTYFPR-UHFFFAOYSA-N
Synonym 2-methyl-5-fluorobenzoic acid, 5-fluoro-o-toluic acid, 3-fluoro-6-methylbenzoic acid, 5-fluoro-2-methylbenzoicacid, benzoic acid, 5-fluoro-2-methyl, 5-fluoro-2-methyl-benzoic acid, 5-fluoro-2-methyl benzoic acid, benzoicacid, 5-fluoro-2-methyl, pubchem1326
PubChem CID 182114
IUPAC Name 5-fluoro-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)F)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1047603 View Documents Alfa Aesar™
A1047603
1g Each for $27.76
 
AAA1047606 View Documents Alfa Aesar™
A1047606
5g Each for $97.61
 
AAA1047614 View Documents Alfa Aesar™
A1047614
25g Each for $479.59
 
Specifications

Chemical Identifiers

33184-16-6
154.14
JVBLXLBINTYFPR-UHFFFAOYSA-N
182114
CC1=C(C=C(C=C1)F)C(=O)O
C8H7FO2
MFCD00042294
2-methyl-5-fluorobenzoic acid, 5-fluoro-o-toluic acid, 3-fluoro-6-methylbenzoic acid, 5-fluoro-2-methylbenzoicacid, benzoic acid, 5-fluoro-2-methyl, 5-fluoro-2-methyl-benzoic acid, 5-fluoro-2-methyl benzoic acid, benzoicacid, 5-fluoro-2-methyl, pubchem1326
5-fluoro-2-methylbenzoic acid

Specifications

130°C to 132°C
33184-16-6
MFCD00042294
2-methyl-5-fluorobenzoic acid, 5-fluoro-o-toluic acid, 3-fluoro-6-methylbenzoic acid, 5-fluoro-2-methylbenzoicacid, benzoic acid, 5-fluoro-2-methyl, 5-fluoro-2-methyl-benzoic acid, 5-fluoro-2-methyl benzoic acid, benzoicacid, 5-fluoro-2-methyl, pubchem1326
CC1=C(C=C(C=C1)F)C(=O)O
154.14
154.14
5-Fluoro-2-methylbenzoic acid
1g
C8H7FO2
2249813
JVBLXLBINTYFPR-UHFFFAOYSA-N
5-fluoro-2-methylbenzoic acid
182114
99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning