Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylbenzeneboronic acid, 99%

$289.30 - $289.30

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylbenzeneboronic acid
CAS 163517-62-2
Molecular Formula C7H8BFO2
Formula Weight 153.95
MDL Number MFCD03095047
Synonym 5-Fluoro-2-methylphenylboronic acid; 5-Fluoro-o-tolylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1981906

  SDS
alfa aesar™
L1981906
5g Each for $289.30
Description & Specifications

Specifications

Formula Weight 153.95
Molecular Formula C7H8BFO2
Synonym 5-Fluoro-2-methylphenylboronic acid; 5-Fluoro-o-tolylboronic acid
Assay 99%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD03095047
Health Hazard 1 Warning
Melting Point 148° to 149°C
Chemical Name or Material 5-Fluoro-2-methylbenzeneboronic acid
CAS 163517-62-2
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.