Please login to your online account to display your discounted pricing

5-Fluoro-2-methylbenzamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylbenzamide
CAS 175278-28-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8FNO
Formula Weight 153.16
MDL Number MFCD00205105
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

TL00714EA

  SDS
fisher scientific company
TL00714EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TL00714DA

  SDS
fisher scientific company
TL00714DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.16
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 175278-28-1
MDL Number MFCD00205105
Assay Percent Range 97%
Chemical Name or Material 5-Fluoro-2-methylbenzamide
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H8FNO
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.