Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methylbenzaldehyde, 97%

$59.89 - $641.88

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methylbenzaldehyde
CAS 22062-53-9
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD02261763
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2679903

 
alfa aesar™
H2679903
1g Each for $59.89

AAH2679906

 
alfa aesar™
H2679906
5g Each for $197.03

AAH2679914

 
alfa aesar™
H2679914
25g Each for $641.88
Description & Specifications

Specifications

Formula Weight 138.14
Health Hazard 1 Warning
Chemical Name or Material 5-Fluoro-2-methylbenzaldehyde
Assay 97%
Molecular Formula C8H7FO
MDL Number MFCD02261763
CAS 22062-53-9
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 77°C (170°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.