missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoro-2-methylaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2538109

 View more versions of this product

Catalog No. AAB2538109



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Fluoro-2-methylaniline
35°C to 38°C
90°C (194°F)
C7H8FN
MFCD00007764
UN2811
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
CC1=CC=C(F)C=C1N
125.15
125.15
367-29-3
98°C to 100°C (15 mmHg)
Characteristic
1.538
10 g
2637584
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
5-fluoro-2-methylaniline
67774
98%

Chemical Identifiers

367-29-3
125.15
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
67774
CC1=CC=C(F)C=C1N
C7H8FN
MFCD00007764
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
5-fluoro-2-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-689-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only