Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-methylacetophenone, 97%

$77.15 - $229.03

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-methylacetophenone
CAS 29427-49-4
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD06660189
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2681903

  SDS
alfa aesar™
H2681903
1g Each for $77.15

AAH2681906

  SDS
alfa aesar™
H2681906
5g Each for $229.03
Description & Specifications

Specifications

Formula Weight 152.17
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 29427-49-4
Health Hazard 1 Warning
Chemical Name or Material 5'-Fluoro-2'-methylacetophenone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD06660189
Molecular Formula C9H9FO
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.