Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methoxypyridine, 97%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methoxypyridine
CAS 51173-04-7
Molecular Formula C6H6FNO
Formula Weight 127.12
MDL Number MFCD03095252
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3099106

  SDS
alfa aesar™
H3099106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3099114

  SDS
alfa aesar™
H3099114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3099103

  SDS
alfa aesar™
H3099103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Fluoro-2-methoxypyridine
Quantity 5g
CAS 51173-04-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Assay 97%
Molecular Formula C6H6FNO
Formula Weight 127.12
MDL Number MFCD03095252
UN Number UN1993
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C