missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methoxypyridine-3-carboxaldehyde, 98%

$38.28 - $452.77

Chemical Identifiers

CAS 351410-62-3
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD04972398
InChI Key YAGPZRLCMRMSFP-UHFFFAOYSA-N
Synonym 2-methoxy-5-fluoropyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy, 3-pyridinecarboxaldehyde,5-fluoro-2-methoxy, 5-fluoro-2-methoxy-3-pyridine carboxaldehyde, 5-fluoro-2-methoxy-3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy-pyridine-3-carbaldehyde, 5-fluoro-2-methoxynicotinaldehyde, 5-fluoro-2-methoxypyridine-3-carboxaldehyde, 5-fluoro-3-formyl-2-methoxypyridine, pubchem16100
PubChem CID 22678323
IUPAC Name 5-fluoro-2-methoxypyridine-3-carbaldehyde
SMILES COC1=NC=C(C=C1C=O)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64696MD View Documents Alfa Aesar™
H64696MD
250mg Each for $38.28
 
AAH6469603 View Documents Alfa Aesar™
H6469603
1g Each for $113.53
 
AAH6469606 View Documents Alfa Aesar™
H6469606
5g Each for $452.77
 
Specifications

Chemical Identifiers

351410-62-3
155.128
YAGPZRLCMRMSFP-UHFFFAOYSA-N
22678323
COC1=NC=C(C=C1C=O)F
C7H6FNO2
MFCD04972398
2-methoxy-5-fluoropyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy, 3-pyridinecarboxaldehyde,5-fluoro-2-methoxy, 5-fluoro-2-methoxy-3-pyridine carboxaldehyde, 5-fluoro-2-methoxy-3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy-pyridine-3-carbaldehyde, 5-fluoro-2-methoxynicotinaldehyde, 5-fluoro-2-methoxypyridine-3-carboxaldehyde, 5-fluoro-3-formyl-2-methoxypyridine, pubchem16100
5-fluoro-2-methoxypyridine-3-carbaldehyde

Specifications

Air and moisture sensitive
351410-62-3
MFCD04972398
2-methoxy-5-fluoropyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy, 3-pyridinecarboxaldehyde,5-fluoro-2-methoxy, 5-fluoro-2-methoxy-3-pyridine carboxaldehyde, 5-fluoro-2-methoxy-3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy-pyridine-3-carbaldehyde, 5-fluoro-2-methoxynicotinaldehyde, 5-fluoro-2-methoxypyridine-3-carboxaldehyde, 5-fluoro-3-formyl-2-methoxypyridine, pubchem16100
COC1=NC=C(C=C1C=O)F
155.128
155.13
98%
250mg
C7H6FNO2
14158282
YAGPZRLCMRMSFP-UHFFFAOYSA-N
5-fluoro-2-methoxypyridine-3-carbaldehyde
22678323
Solid
5-Fluoro-2-methoxypyridine-3-carboxaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon