missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoro-2-methoxyphenylboronic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393890050

 View more versions of this product

Catalog No. AC393890050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

5-Fluoro-2-methoxyphenylboronic acid
5g
97.5
Authentic
Crystalline Powder
98%
169.95
White
179897-94-0
100.0
Glass bottle
≥96.0%
C7H8BFO3
MFCD01863526

Chemical Identifiers

179897-94-0
100.0
C7H8BFO3
MFCD01863526
≥96.0%
97.5
98%
169.95
5-Fluoro-2-methoxyphenylboronic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.