Please login to your online account to display your discounted pricing

5-Fluoro-2-methoxyphenylboronic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methoxyphenylboronic acid
CAS 179897-94-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 98%
Molecular Formula C7H8BFO3
Formula Weight 169.95
MDL Number MFCD01863526
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393890010

  SDS
thermo fisher scientific
AC393890010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393890050

  SDS
thermo fisher scientific
AC393890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 169.95
Quantity 1g
Color White
CAS Max % 100.0
Chemical Name or Material 5-Fluoro-2-methoxyphenylboronic acid
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
CAS Min % 97.5
Molecular Formula C7H8BFO3
MDL Number MFCD01863526
CAS 179897-94-0
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.