Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methoxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Formula Weight 252.09
Synonym 5-Fluoro-2-methoxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6270103

 
alfa aesar™
H6270103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62701MD

 
alfa aesar™
H62701MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.09
Chemical Name or Material 5-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Synonym 5-Fluoro-2-methoxyphenylboronic acid pinacol ester
Quantity 1g