Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-methoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-methoxyaniline
CAS 1978-39-8
Molecular Formula C7H8FNO
Formula Weight 141.15
MDL Number MFCD00077537
UN Number UN2810
Synonym 5-Fluoro-o-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3252503

 
alfa aesar™
H3252503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32525MD

 
alfa aesar™
H32525MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.15
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Chemical Name or Material 5-Fluoro-2-methoxyaniline
Assay 97%
Molecular Formula C7H8FNO
MDL Number MFCD00077537
CAS 1978-39-8
UN Number UN2810
Synonym 5-Fluoro-o-anisidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.