Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-methoxyacetophenone, 99%

$33.72 - $78.19

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-methoxyacetophenone
CAS 445-82-9
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD00671763
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2610803

 
alfa aesar™
H2610803
1g Each for $78.19

AAH26108MD

 
alfa aesar™
H26108MD
250mg Each for $33.72
Description & Specifications

Specifications

Formula Weight 168.17
Chemical Name or Material 5'-Fluoro-2'-methoxyacetophenone
Assay 99%
Molecular Formula C9H9FO2
MDL Number MFCD00671763
CAS 445-82-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.