Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-iodoacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-iodoacetophenone
CAS 914225-70-0
Formula Weight 264.04
MDL Number MFCD09033159
Beilstein 22044599
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6647503

 
alfa aesar™
H6647503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6647506

 
alfa aesar™
H6647506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66475MD

 
alfa aesar™
H66475MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 264.04
Assay 97%
MDL Number MFCD09033159
CAS 914225-70-0
Beilstein 22044599
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Liquid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Light Sensitive
Color Brown
Quantity 1g
Chemical Name or Material 5'-Fluoro-2'-iodoacetophenone

Applications
5′-Fluoro-2′-iodoacetophenone is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.