Please login to your online account to display your discounted pricing

5-Fluoro-2-hydroxypyridine, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC432580250

 View more versions of this product

Catalog No. AC432580250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 113.09
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 25g
Color White to off-white
Melting Point 151° to 155°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
If medical advice is needed, have product container or label at hand.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 51173-05-8
Chemical Name or Material 5-Fluoro-2-hydroxypyridine
MDL Number MFCD03092918
Assay Percent Range 97%
Molecular Formula C5H4FNO
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Fluoro-2-hydroxypyridine
CAS 51173-05-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD03092918
Accessories

GHS P Statement:
If medical advice is needed, have product container or label at hand.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents