Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-hydroxybenzaldehyde, 98+%

$156.70 - $156.70

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-hydroxybenzaldehyde
CAS 347-54-6
Molecular Formula C7H5FO2
Formula Weight 140.11
MDL Number MFCD01090997
Synonym 5-Fluorosalicylaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2572403

  SDS
alfa aesar™
B2572403
1g Each for $156.70
Description & Specifications

Specifications

Formula Weight 140.11
MDL Number MFCD01090997
Health Hazard 1 Warning
Quantity 1g
Synonym 5-Fluorosalicylaldehyde
Melting Point 82° to 85°C
Chemical Name or Material 5-Fluoro-2-hydroxybenzaldehyde
CAS 347-54-6
Molecular Formula C7H5FO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.