Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-hydroxyacetophenone, 98%

$111.95 - $453.54

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-hydroxyacetophenone
CAS 394-32-1
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD00011668
Beilstein 2359087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1561406

  SDS
alfa aesar™
A1561406
5g Each for $111.95

AAA1561414

  SDS
alfa aesar™
A1561414
25g Each for $453.54
Description & Specifications

Specifications

Formula Weight 154.14
Boiling Point 65° to 66°C (8mmHg)
Quantity 5g
Chemical Name or Material 5'-Fluoro-2'-hydroxyacetophenone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 56° to 58°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
RTECS Number AM8502300
MDL Number MFCD00011668
Beilstein 2359087
CAS 394-32-1
Health Hazard 1 Warning
Molecular Formula C8H7FO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.