missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Fluoro-2'-hydroxyacetophenone, 98%

Manufacturer:  Alfa Aesar™ A1561414

 View more versions of this product

Catalog No. AAA1561414



Specifications

Specifications

5'-Fluoro-2'-hydroxyacetophenone
56°C to 58°C
394-32-1
MFCD00011668
5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone
CC(=O)C1=C(C=CC(=C1)F)O
154.14
154.14
65°C to 66°C (8mmHg)
25g
C8H7FO2
2359087
KOFFXZYMDLWRHX-UHFFFAOYSA-N
1-(5-fluoro-2-hydroxyphenyl)ethanone
95992
98%

Chemical Identifiers

394-32-1
154.14
KOFFXZYMDLWRHX-UHFFFAOYSA-N
95992
CC(=O)C1=C(C=CC(=C1)F)O
C8H7FO2
MFCD00011668
5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone
1-(5-fluoro-2-hydroxyphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AM8502300

Documents