Please login to your online account to display your discounted pricing

5'-Fluoro-2'-hydroxyacetophenone, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5'-Fluoro-2'-hydroxyacetophenone
Name Note 97%
CAS 394-32-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7FO2
Linear Formula FC6H3(OH)COCH3
Formula Weight 154.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC277930050

  SDS
thermo fisher scientific
AC277930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystals or Crystalline Powder
Formula Weight 154.14
Quantity 5g
Color Light yellow to beige-brown
EINECS Number 206-893-5
RTECS Number AM8502300
Melting Point 54° to 59°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 5'-Fluoro-2'-hydroxyacetophenone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H7FO2
Linear Formula FC6H3(OH)COCH3
MDL Number MFCD00011668
CAS 394-32-1
Synonym 1-(5-Fluoro-2-hydroxyphenyl)-1-ethanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.