missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-hydroxy-3-nitropyridine, 98%

$33.01 - $476.51

Chemical Identifiers

CAS 136888-20-5
Molecular Formula C5H3FN2O3
Molecular Weight (g/mol) 158.088
MDL Number MFCD05662412
InChI Key BLFUHTOZIOQGBU-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxy-3-nitropyridine, 5-fluoro-3-nitropyridin-2-ol, 5-fluoro-3-nitropyridin-2 1h-one, 2-hydroxy-3-nitro-5-fluoropyridine, 2-hydroxy-5-fluoro-3-nitropyridine, 2 1h-pyridinone, 5-fluoro-3-nitro, pubchem2975, acmc-1cgzr, ksc520k3n, 5-fluoro-2-hydroxy-3-nitrop
PubChem CID 10583047
IUPAC Name 5-fluoro-3-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64441MD View Documents Alfa Aesar™
H64441MD
250mg Each for $33.01
 
AAH6444103 View Documents Alfa Aesar™
H6444103
1g Each for $104.55
 
AAH6444106 View Documents Alfa Aesar™
H6444106
5g Each for $476.51
 
Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

136888-20-5
158.088
BLFUHTOZIOQGBU-UHFFFAOYSA-N
10583047
C1=C(C(=O)NC=C1F)[N+](=O)[O-]
C5H3FN2O3
MFCD05662412
5-fluoro-2-hydroxy-3-nitropyridine, 5-fluoro-3-nitropyridin-2-ol, 5-fluoro-3-nitropyridin-2 1h-one, 2-hydroxy-3-nitro-5-fluoropyridine, 2-hydroxy-5-fluoro-3-nitropyridine, 2 1h-pyridinone, 5-fluoro-3-nitro, pubchem2975, acmc-1cgzr, ksc520k3n, 5-fluoro-2-hydroxy-3-nitrop
5-fluoro-3-nitro-1H-pyridin-2-one

Specifications

250mg
C5H3FN2O3
7582013
BLFUHTOZIOQGBU-UHFFFAOYSA-N
5-fluoro-3-nitro-1H-pyridin-2-one
10583047
Solid
5-Fluoro-2-hydroxy-3-nitropyridine
136888-20-5
MFCD05662412
5-fluoro-2-hydroxy-3-nitropyridine, 5-fluoro-3-nitropyridin-2-ol, 5-fluoro-3-nitropyridin-2 1h-one, 2-hydroxy-3-nitro-5-fluoropyridine, 2-hydroxy-5-fluoro-3-nitropyridine, 2 1h-pyridinone, 5-fluoro-3-nitro, pubchem2975, acmc-1cgzr, ksc520k3n, 5-fluoro-2-hydroxy-3-nitrop
C1=C(C(=O)NC=C1F)[N+](=O)[O-]
158.088
158.09
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning