Learn More
5-Fluoro-2'-deoxyuridine, 98+%, Thermo Scientific Chemicals
$197.62 - $639.16
Chemical Identifiers
CAS | 50-91-9 |
---|---|
Molecular Formula | C9H11FN2O5 |
Molecular Weight (g/mol) | 246.19 |
MDL Number | MFCD00006530 |
InChI Key | ODKNJVUHOIMIIZ-RRKCRQDMSA-N |
Synonym | floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd |
PubChem CID | 5790 |
ChEBI | CHEBI:60761 |
IUPAC Name | 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL16497MC
|
Thermo Scientific Chemicals
L16497MC |
100 mg |
Each for $197.62
|
|
|||||
AAL16497ME
|
Thermo Scientific Chemicals
L16497ME |
500 mg |
Each for $639.16
|
|
|||||
Description
5-Fluoro-2'-deoxyuridine acts as an antiviral and antineoplastic agent. It is also employed for renal function diagnosis. Further, it is used as an experimental anticancer agent, which shows activity against a variety of malignant neoplasms. It is also used as an inhibitor of thymidylate synthetase and DNA synthesis. In addition to this, it plays an important role in the treatment of colorectal cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, alcohol, isopropyl alcohol, methanol, aqueous buffers, acetone, dimethyl sulfoxide, and ethanol. Insoluble in ether, benzene and chloroform.
Notes
Store in a cool place.Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
50-91-9 | |
246.19 | |
ODKNJVUHOIMIIZ-RRKCRQDMSA-N | |
5790 | |
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
C9H11FN2O5 | |
MFCD00006530 | |
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd | |
CHEBI:60761 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O |
Specifications
50-91-9 | |
C9H11FN2O5 | |
100 mg | |
2645818 | |
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd | |
ODKNJVUHOIMIIZ-RRKCRQDMSA-N | |
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
5790 | |
246.2 | |
5-Fluoro-2'-deoxyuridine |
148°C to 151°C | |
MFCD00006530 | |
UN2811 | |
14,4112 | |
Soluble in water,alcohol,isopropyl alcohol,methanol,aqueous buffers,acetone,dimethyl sulfoxide,and ethanol. Insoluble in ether,benzene and chloroform. | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O | |
246.19 | |
CHEBI:60761 | |
≥98% |
Safety and Handling
GHS H Statement
H360-H341-H302
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
P201-P202-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H301-H315-H319-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-072-5
RTECSNumber : YU7535000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only