Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2,4-dichloropyrimidine, 98%

$28.93 - $28.93

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine
CAS 2927-71-1
Molecular Formula C4HCl2FN2
Formula Weight 166.97
MDL Number MFCD00233551
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2628803

  SDS
alfa aesar™
H2628803
1g Each for $28.93

AAH2628806

  SDS
alfa aesar™
H2628806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.97
Melting Point 36° to 39°C
CAS 2927-71-1
Flash Point 106°C (223°F)
Assay 98%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Boiling Point 80°C (16mmHg)
UN Number UN3261
Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine
Quantity 1g
Molecular Formula C4HCl2FN2
MDL Number MFCD00233551
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.