missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-1-indanone, 99%

$59.53 - $210.92

Chemical Identifiers

CAS 700-84-5
Molecular Formula C9H7FO
Formula Weight 150.15
MDL Number MFCD00041031
Beilstein 1617799
Chemical Name or Material 5-Fluoro-1-indanone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1094903 SDS Alfa Aesar™
L1094903
1g Each for $59.53
AAL1094906 SDS Alfa Aesar™
L1094906
5g Each for $210.92
Specifications Description & Specifications

Chemical Identifiers

700-84-5
150.15
1617799
C9H7FO
MFCD00041031
5-Fluoro-1-indanone

Specifications

109°C (228°F)
37° to 40°C
700-84-5
150.15
1617799
99%
92° to 93°C (3mmHg)
1g
C9H7FO
MFCD00041031
5-Fluoro-1-indanone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning