missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoro-1-indanone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC271110010

 View more versions of this product

Catalog No. AC271110010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Fluoro-1-indanone
Authentic
92°C to 93°C (3.0mmHg)
35°C to 39°C
98%
97.5
98%
MFCD00041031
Solubility in water: insoluble
C1CC(=O)C2=C1C=C(C=C2)F
150.15
150.15
98%
109°C
Glass bottle
White to Yellow
1g
700-84-5
100.0
C9H7FO
1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-flouro-1-indanone, 5-fluoro indanone, 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 5-fluoroindanone, pubchem13785, pubchem2388
WVPPBVAMKNQXJA-UHFFFAOYSA-N
5-fluoro-2,3-dihydroinden-1-one
136537
Crystalline Low Melting Solid

Chemical Identifiers

700-84-5
150.15
WVPPBVAMKNQXJA-UHFFFAOYSA-N
136537
C1CC(=O)C2=C1C=C(C=C2)F
C9H7FO
MFCD00041031
1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-flouro-1-indanone, 5-fluoro indanone, 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 5-fluoroindanone, pubchem13785, pubchem2388
5-fluoro-2,3-dihydroinden-1-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents