missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethynyl-2′-deoxyuridine, TCI America™

Manufacturer:  TCI America E1057200MG

 View more versions of this product

Catalog No. E1057200MG



Specifications

Specifications

5-Ethynyl-2′-deoxyuridine
199°C
61135-33-9
MFCD01675687
CDEURGJCGCHYFH-DJLDLDEBSA-N
5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
472172
Crystal-Powder at 20°C
White-Yellow
200mg
C11H12N2O5
EdU
C#CC1=CN(C(=O)NC1=O)C2CC(C(O2)CO)O
252.226
252.23
≥98.0% (HPLC,T)

Chemical Identifiers

61135-33-9
252.226
CDEURGJCGCHYFH-DJLDLDEBSA-N
472172
C#CC1=CN(C(=O)NC1=O)C2CC(C(O2)CO)O
C11H12N2O5
MFCD01675687
EdU
5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

RTECSNumber : YU7510000

TSCA : No

Documents