missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethynyl-2′-deoxycytidine, TCI America™

Manufacturer:  TCI America E109350MG

 View more versions of this product

Catalog No. E109350MG



Specifications

Specifications

5-Ethynyl-2′-deoxycytidine
190°C
69075-47-4
MFCD22417249
HMJSYXIPNSASJY-IWSPIJDZSA-N
4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
90176383
Crystal-Powder at 20°C
Undesignated
50mg
C11H13N3O4
EdC
C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O
251.242
251.24
≥98.0% (N)

Chemical Identifiers

69075-47-4
251.242
HMJSYXIPNSASJY-IWSPIJDZSA-N
90176383
C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O
C11H13N3O4
MFCD22417249
EdC
4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents