Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethyluracil, 98+%

$83.18 - $83.18

Chemical Identifiers

Chemical Name or Material 5-Ethyluracil
CAS 4212-49-1
Molecular Formula C6H8N2O2
Formula Weight 140.14
MDL Number MFCD00079187
Beilstein 119494
Synonym 2,4-Dihydroxy-5-ethylpyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL10861MD

  SDS
alfa aesar™
L10861MD
250mg Each for $83.18
Description & Specifications

Specifications

Formula Weight 140.14
Chemical Name or Material 5-Ethyluracil
Molecular Formula C6H8N2O2
MDL Number MFCD00079187
CAS 4212-49-1
Assay ≥98%
Beilstein 119494
Synonym 2,4-Dihydroxy-5-ethylpyrimidine
Melting Point 308° to 310°C
Quantity 250mg