Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethylpyridine-2,3-dicarboxylic acid, 94%

$238.07 - $238.07

Chemical Identifiers

Chemical Name or Material 5-Ethylpyridine-2,3-dicarboxylic acid
CAS 102268-15-5
Molecular Formula C9H9NO4
Formula Weight 195.18
MDL Number MFCD00071733
Synonym 5-Ethylquinolinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945114

  SDS
alfa aesar™
L1945114
25g Each for $238.07
Description & Specifications

Specifications

Formula Weight 195.18
Chemical Name or Material 5-Ethylpyridine-2,3-dicarboxylic acid
MDL Number MFCD00071733
CAS 102268-15-5
Health Hazard 1 Warning
Assay 94%
Molecular Formula C9H9NO4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Synonym 5-Ethylquinolinic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 152° to 154°C
Quantity 25g
RTECS Number US8005000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.