missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol

Manufacturer:  Alfa Aesar™ 43972AC

 View more versions of this product

Catalog No. AA43972AC



Specifications

Specifications

5-Ethylidene-2-norbornene
38°C (101°F)
25mL
16219-75-3
MFCD00167576
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
CC=C1CC2CC1C=C2
120.195
120.19
98%
0.89g/mL
146°C
endo + exo, Stabilized with 100ppm 4-tert-butylcatechol
C9H12
UN3295
OJOWICOBYCXEKR-WHZSQGQPSA-N
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
12456033
Liquid

Chemical Identifiers

16219-75-3
120.195
OJOWICOBYCXEKR-WHZSQGQPSA-N
12456033
CC=C1CC2CC1C=C2
C9H12
MFCD00167576
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : RB9450000

TSCA : TSCA

Documents