Learn More
5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043972AP
Description
5-Ethylidene-2-norbornene is used as a self healing agent for polymeric composites. It is also used to reinforce multiwalled carbon nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Ethylidene-2-norbornene | |
16219-75-3 | |
146°C | |
C9H12 | |
500 mL | |
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component | |
OJOWICOBYCXEKR-WHZSQGQPSA-N | |
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene | |
12456033 | |
98% |
endo + exo, Stabilized with 100ppm 4-tert-butylcatechol | |
0.89 g/mL | |
38°C (101°F) | |
MFCD00167576 | |
UN3295 | |
Soluble in water (0.01% at 20C). | |
CC=C1CC2CC1C=C2 | |
120.195 | |
120.19 | |
Liquid |
Chemical Identifiers
16219-75-3 | |
120.195 | |
OJOWICOBYCXEKR-WHZSQGQPSA-N | |
12456033 | |
CC=C1CC2CC1C=C2 |
C9H12 | |
MFCD00167576 | |
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component | |
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene |
Safety and Handling
GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P330-P332+P313-P363-P370+P378q-P501c
H226-H300-H315-H319-H331
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 240-347-7
RTECSNumber : RB9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only