Please login to your online account to display your discounted pricing

5-Ethylidene-2-norbornene, 98+%, mixt. of endo- and exo-isomers, stabilized, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Ethylidene-2-norbornene
Name Note 98+%
CAS 16219-75-3
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H12
Formula Weight 120.19
MDL Number MFCD00167576
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166010050

  SDS
thermo fisher scientific
166010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.19
Physical Form Liquid
Assay ≥98.0%
Quantity 5g
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 240-347-7
RTECS Number RB9450000
Density 0.8900g/mL
CAS Max % 100.0
CAS Min % 98.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Stabilizer 100ppm p-tert.-butylcatechol
Refractive Index 1.4895 to 1.4915 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98+%
Chemical Name or Material 5-Ethylidene-2-norbornene
Molecular Formula C9H12
CAS 16219-75-3
MDL Number MFCD00167576
Assay Percent Range 98+%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.