missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone, TCI America™

Manufacturer:  TCI America E080025G

 View more versions of this product

Catalog No. E080025G



Specifications

Specifications

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone
White-Yellow
698-10-2
MFCD00036673
IUFQZPBIRYFPFD-UHFFFAOYSA-N
2-ethyl-4-hydroxy-3-methyl-2H-furan-5-one
61199
142.15
≥97.0% (GC,T)
94°C
25g
C7H10O3
5-ethyl-3-hydroxy-4-methyl-2 5h-furanone, 5-ethyl-3-hydroxy-4-methylfuran-2 5h-one, 2 5h-furanone, 5-ethyl-3-hydroxy-4-methyl, 3-hydroxy-4-methyl-5-ethyl-2-furanone, fema no. 3153, 4-methyl-5-ethyl-3-hydroxyfuranone, ethyl fenugreek lactone, 3-hydroxy-4-methyl-5-ethyl-2 5h furanone, 5-ethyl-3-hydroxy-4-methyl-2 5h furanone, 2-hydroxy-3-methyl-gamma-2-hexenolactone
CCC1C(=C(C(=O)O1)O)C
142.154
CHEBI:67940
Crystal-Lump at 20°C

Chemical Identifiers

698-10-2
142.154
IUFQZPBIRYFPFD-UHFFFAOYSA-N
61199
2-ethyl-4-hydroxy-3-methyl-2H-furan-5-one
C7H10O3
MFCD00036673
5-ethyl-3-hydroxy-4-methyl-2 5h-furanone, 5-ethyl-3-hydroxy-4-methylfuran-2 5h-one, 2 5h-furanone, 5-ethyl-3-hydroxy-4-methyl, 3-hydroxy-4-methyl-5-ethyl-2-furanone, fema no. 3153, 4-methyl-5-ethyl-3-hydroxyfuranone, ethyl fenugreek lactone, 3-hydroxy-4-methyl-5-ethyl-2 5h furanone, 5-ethyl-3-hydroxy-4-methyl-2 5h furanone, 2-hydroxy-3-methyl-gamma-2-hexenolactone
CHEBI:67940
CCC1C(=C(C(=O)O1)O)C
Safety and Handling

Safety and Handling

TSCA : Yes

Documents