Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Ethyl-2-thiophenecarboxaldehyde, 98%

$56.96 - $785.03

Chemical Identifiers

Chemical Name or Material 5-Ethyl-2-thiophenecarboxaldehyde
CAS 36880-33-8
Molecular Formula C7H8OS
Formula Weight 140.21
MDL Number MFCD00143457
Beilstein 108539
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2371103

 
alfa aesar™
B2371103
1g Each for $56.96

AAB2371106

 
alfa aesar™
B2371106
5g Each for $192.41

AAB2371114

 
alfa aesar™
B2371114
25g Each for $785.03
Description & Specifications

Specifications

Formula Weight 140.21
Chemical Name or Material 5-Ethyl-2-thiophenecarboxaldehyde
TSCA TSCA
Assay 98%
Molecular Formula C7H8OS
MDL Number MFCD00143457
CAS 36880-33-8
Beilstein 108539
Boiling Point 65°C (2mmHg)
Quantity 1g
Flash Point 109°C (228°F)