missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethyl-2-methylpyridine, TCI America™

Manufacturer:  TCI America E014225ML

 View more versions of this product

Catalog No. E014225ML



Specifications

Specifications

5-Ethyl-2-methylpyridine
Yellow
104-90-5
MFCD00006344
aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine
CCC1=CN=C(C=C1)C
121.183
121.18
≥95.0% (GC)
178°C
25mL
C8H11N
2300
NTSLROIKFLNUIJ-UHFFFAOYSA-N
5-ethyl-2-methylpyridine
7728
Clear Liquid at 20°C

Chemical Identifiers

104-90-5
121.183
NTSLROIKFLNUIJ-UHFFFAOYSA-N
7728
CCC1=CN=C(C=C1)C
C8H11N
MFCD00006344
aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine
5-ethyl-2-methylpyridine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0714

RTECSNumber : TJ6825000

TSCA : Yes

Documents